- Pk:96
- Primary antibody reactivity:Human
- Description:Human TSLP ELISA Kit
- Detection range:31,2 - 2000 pg/ml
- Sensitivity:<10 pg/ml
Human TSLP ELISA kit for the quantitative detection of human thymic stromal lymphopoietin (TSLP) in cell culture supernates, serum and plasma (heparin, EDTA).
TSLP is a protein belonging to the cytokine family. This gene is mapped to 5q22.1. It encodes a hemopoietic cytokine proposed to signal through a heterodimeric receptor complex composed of the thymic stromal lymphopoietin receptor and the IL-7R alpha chain. It mainly impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c(+) dendritic cells. The protein promotes T helper type 2 (TH2) cell responses that are associated with immunity in various inflammatory diseases, including asthma, allergic inflammation and chronic obstructive pulmonary disease. The protein is therefore considered a potential therapeutic target for the treatment of such diseases.