Order Entry
Norway
ContactUsLinkComponent
Ion chromatography columns, transgenomic
Ion chromatography columns, transgenomic
  554-4713
Ion chromatography columns, transgenomic
  554-4713

 

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Description:
    ICSep ION-120
  • Length:
    120 mm
  • Seq:
    01
  • Pk:
    1