LC3 (from E. coli)
Marketsource
Supplier: Biorbyt
Om dette produktet
2 Alternativer tilgjenglige nedenfor
Product Details & Documents
Documents
orb81209
Datablad
Specifications for Product Family
Spesifikasjoner
Spesifikasjoner
- Catalog No:
- BIRBORB81209-5
- BIRBORB81209-20
- Protein/peptide name:
- LC3
- LC3
- Protein synonyms:
- ATG8F
- ATG8F
- Source:
- E. coli
- E. coli
- Sequence:
- MPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVSVSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH.
- MPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVSVSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH.
- Purity:
- Greater than 95.0% as determined by SDS-PAGE.
- Greater than 95.0% as determined by SDS-PAGE.
- Formulation:
- 1mg/ml solution containing 20 mM Tris pH-8, 1mM DTT, 0.1M NaCl 10% glycerol.
- 1mg/ml solution containing 20 mM Tris pH-8, 1mM DTT, 0.1M NaCl 10% glycerol.
- Storage conditions:
- MAP1LC3A Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
- MAP1LC3A Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
- Tested applications:
- SDS-PAGE
- SDS-PAGE
Spesifikasjoner
Product Family Options
Produktinformasjon
- PkAvailabilityPrice
- Spesifikasjoner:Marketsource
More
Specifications
Cat. No.BIRBORB81209-5Protein/peptide nameLC3Protein synonymsATG8FSourceE. coliSequenceMPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVSVSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH.PurityGreater than 95.0% as determined by SDS-PAGE.Formulation1mg/ml solution containing 20 mM Tris pH-8, 1mM DTT, 0.1M NaCl 10% glycerol.Storage conditionsMAP1LC3A Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.Tested applicationsSDS-PAGEMarketsource - Spesifikasjoner:Marketsource
More
Specifications
Cat. No.BIRBORB81209-20Protein/peptide nameLC3Protein synonymsATG8FSourceE. coliSequenceMPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVSVSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH.PurityGreater than 95.0% as determined by SDS-PAGE.Formulation1mg/ml solution containing 20 mM Tris pH-8, 1mM DTT, 0.1M NaCl 10% glycerol.Storage conditionsMAP1LC3A Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.Tested applicationsSDS-PAGEMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



