Rat recombinant RAP (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Lyophilized RAP Rat although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution RAP Rat should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Receptor Associated
- Protein/peptide name:RAP
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized RAP Rat in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:The protein (1mg/ml) was lyophilized after from sterile solution containing TBS pH-7.5, 0.1% BSA and 0.09% NaN3.
- Tested applications:SDS-PAGE
Frequently Bought Together