Human Recombinant GUCA2B (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated thaw-freeze cycles and store at −80 °C. Reconstituted protein can be stored at 4 °C for limited period of time.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:Guanylate cyclase activator 2B
- Protein/peptide name:GUCA2B
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE Three-step procedure using IMAC and size exclusion chromatography before and after refolding.
- Solubility:Add 0.2 uml of deionized H2O and let the lyophilized pellet dissolve completely.
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in 50mM Phosphate buffer pH-7.4.
- Tested applications:SDS-PAGE
Frequently Bought Together