Order Entry
Norway
ContactUsLinkComponent
 
Human Galectin 8 (from E. coli)
 :  Biorbyt
 
Human Galectin 8 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    LGALS8 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles .
  • Protein synonyms:
    Gal-8
  • Protein/peptide name:
    Galectin 8
  • Purity:
    Greater than 90.0% as determined by SDS-PAGE.
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH MMLSLNNLQN IIYNPVIPFV GTIPDQLDPG TLIVIRGHVP SDADRFQVDL QNGSSMKPRA DVAFHFNPRF KRAGCIVCNT LINEKWGREE ITYDTPFKRE KSFEIVIMVL KDKFQVAVNG KHTLLYGHRI GPEKIDTLGI YGKVNIHSIG FSFSSDLQST QASSLELTEI SRENVPKSGT PQLRLPFAAR LNTPMGPGRT VVVKGEVNAN AKSFNVDLLA GKSKDIALHL NPRLNIKAFV RNSFLQESWG EEERNITSFP FSPGMYFEMI IYCDVREFKV AVNGVHSLEY KHRFKELSSI DTLEINGDIH LLEVRSW.
  • Formulation:
    The Galectin-8 His tag 0.5mg/ml protein solution contains 20 mM Tris-HCl pH-8, 0.1M NaCl, 10% glycerol 1mM DTT.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together