Human recombinant Galectin 3 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized Galectin-3 Recombinant although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Galectin-3 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:Determined by its ability to chemoattract human blood monocytes. Chemotactic activity was observed at concentration of ug/ml with peak response obtained at 220 ug/ml.
- Protein synonyms:CBP 35
- Protein/peptide name:Galectin 3
- Purity:Greater than 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized LGALS3 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:ADNFSLHDAL SGSGNPNPQG WPGAWGNQPA GAGGYPGASY PGAYPGQAPP GAYPGQAPPG AYHGAPGAYP GAPAPGVYPG PPSGPGAYPS SGQPSAPGAY PATGPYGAPA GPLIVPYNLP LPGGVVPRML ITILGTVKPN ANRIALDFQR GNDVAFHFNP RFNENNRRVI VCNTKLDNNW GREERQSVFP FESGKPFKIQ VLVEPDHFKV AVNDAHLLQY NHRVKKLNEI SKLGISGDID LTSASYTMI.
- Formulation:The Galectin-3 protein was lyophilized from concentrated (1mg/ml) containing 10mM sodium phosphate buffer, pH-7.5 and 50mM NaCl.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together