Human adiponectin (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized Acrp30 Human at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted Acrp30 Human can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized Acrp30 Human remains stable until the expiry date when stored at −20 °C.
- Gene ID:9370
- Protein synonyms:ACDC
- Protein/peptide name:Adiponectin
- Purity:Acrp30 Human purity is greater than 95% as determined by SDS-PAGE.
- Solubility:It is recommended to add deionized water to prepare working stock solution of approximately 0.5mg/ml and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by an appropriate sterile filter before using it on
- Formulation:Acrp30 Human was filtered (0.4 um) and lyophilized from 0.5 mg/ml in 0.02M Tris buffer pH7.5, 0.15M NaCl.
- Tested applications:SDS-PAGE
Frequently Bought Together