Human recombinant ATG3 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:UbiquitinLikeConjugating Enzyme ATG3
- UniProtKB:Q9NT62 (Human)
- Protein/peptide name:ATG3
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MASMTGGQQMGRGSMQNVINTVKGKALEVAEYLTPVLKESKFKETGVITPEEFVAAGDHLVHHCP TWQWATGEELKVKAYLPTGKQFLVTKNVPCYKRCKQMEYSDELEAIIEEDDGDGGWVDTYHNTGI TGITEAVKEITLENKDNIRLQDCSALCEEEEDEDEGEAADMEEYEESGLLETDEATLDTRKIVEA CKAKTDAGGEDAILQTRTYDLYITYDKYYQTPRLWLFGYDEQRQPLTVEHMYEDISQDHVKKTVT IENHPHLPPPPMCSVHPCRHAEVMKKIIETVAEGGGELGVHLEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2.
- Tested applications:SDS-PAGE
Frequently Bought Together