Order Entry
Norway
ContactUsLinkComponent
Recombinant HIV-1 gp160
Recombinant HIV-1 gp160
Catalog # BIRBORB98895-1
Supplier:  Biorbyt
CAS Number:  
Recombinant HIV-1 gp160
Catalog # BIRBORB98895-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    HIV-1 gp160
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA , WB , IHC-P , RIA

Specifications

About this item