Human recombinant Cystatin C (from Urine)
Catalog # BIRBORB81876-1
Leverandør: Biorbyt
CAS-nummer:
Spesifikasjoner
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:Urine
- Species:Human
- Protein synonyms:AD8
- Protein/peptide name:Cystatin C
- Purity:Purified.
- Formulation:Preservative: 0.09% Sodium Azide