Order Entry
Norway
ContactUsLinkComponent
Mouse TNF alpha (from E. coli)
Mouse TNF alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse TNF alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin is 0.1ng/ml, corresponding to Specific Activity of 10,000,000 Units/mg.
  • Protein synonyms:
    APC1
  • Protein/peptide name:
    TNF alpha
  • Purity:
    > 98.0% as determined by RP-HPLC and SDS-PAGE analysis
  • Solubility:
    It is recommended to reconstitute the lyophilized Tumor Necrosis Factor-alpha in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
  • Formulation:
    Lyophilized from 0.2 um filtered concentrated solution in PBS, pH 7.2.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together