Order Entry
Norway
ContactUsLinkComponent
Mouse Eotaxin 2 (from E. coli)
Mouse Eotaxin 2 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse Eotaxin 2 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    Determined by its ability to chemoattract murine lymphocytes using concentration range of 10ng/ml -100ng/ml.
  • Protein synonyms:
    Small-inducible cytokine A24
  • Protein/peptide name:
    Eotaxin 2
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized CCL24 Mouse Recombinant in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    VTIPSSCCTS FISKKIPEN R VVSYQLAN GS ICPKAGV IFI TKKGHK ICTD PKLLW VQRHI QKLD AKKNQP SKG AKAVRTK FA VQRRRGNS T EV.
  • Formulation:
    The CCL24 protein was lyophilized from concentrated (1mg/ml) sterile solution containing 20 mM Phosphate buffer pH-7.4 and 0.15M sodium chloride.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together