Order Entry
Norway
ContactUsLinkComponent
Human Vimentin (from E. coli)
Human Vimentin (from E. coli)
Supplier:  Biorbyt
Certificates
Human Vimentin (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Avoid multiple freeze-thaw cycles.
  • Protein synonyms:
    CTRCT30
  • Protein/peptide name:
    Vimentin
  • Purity:
    Greater than 95.0% as determined by SDS-gelelctrophoresis.
  • Formulation:
    The Vimentin protein solution contains 50mM Tris-Acetate, pH-7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together