Human recombinant D4GDI (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Protein synonyms:D4-GDI
- Protein/peptide name:D4GDI
- Purity:Greater than 90% as determined by SDS-PAGE.
- Formulation:D4-GDI protein in 50mM Tris-acetate, pH7.5, 20% glycerol and 1mM EDTA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together