Order Entry
Norway
ContactUsLinkComponent
Human recombinant HSP20 (from E. coli)
Human recombinant HSP20 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant HSP20 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    HSP20 although stable at 10 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Heat Shock 20
  • Protein/peptide name:
    HSP20
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Formulation:
    HSP20 lyophilized from 20 mM Tris-acetate, pH-7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF and 15mM beta mercaptoethanol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together