Order Entry
Norway
ContactUsLinkComponent
Anti-Cytokeratin Mouse Monoclonal Antibody (Biotin) [clone: KRTL/1077 KRTH/1076]
Anti-Cytokeratin Mouse Monoclonal Antibody (Biotin) [clone: KRTL/1077 KRTH/1076]
Supplier:  Biotium
Certificates
Anti-Cytokeratin Mouse Monoclonal Antibody (Biotin) [clone: KRTL/1077 KRTH/1076]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB1100-500
  • BNCB1100-100
  • Antigen name:
  • Cytokeratin
  • Cytokeratin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone KRTL/1077 KRTH/1076
  • Clone KRTL/1077 KRTH/1076
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG
  • IgG
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications