Order Entry
Norway
ContactUsLinkComponent
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF594) [clone: 246-B8.E7]
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF594) [clone: 246-B8.E7]
Supplier:  Biotium
Certificates
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF594) [clone: 246-B8.E7]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC941054-500
  • BNC941054-100
  • Antigen symbol:
  • MHC I
  • MHC I
  • Antigen name:
  • MHC Class 1
  • MHC Class 1
  • Antigen synonyms:
  • HLA-B|HLA-A|HLA-C
  • HLA-B|HLA-A|HLA-C
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone 246-B8.E7
  • Clone 246-B8.E7
  • Reactivity:
  • Primates, Human, Mouse
  • Primates, Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 181244; 654404; 77961
  • 181244; 654404; 77961
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications