About this item
Recognizes an 85-115 kDa protein (variation with cell type), identified as intercellular adhesion molecule (ICAM-1) (Workshop IV). It has 7 potential N-linked glycosylation sites. ICAM-1 is a single chain glycoprotein of Ig supergene family, present on unstimulated endothelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1). This interaction enhances antigen-specific T-cell activation. ICAM-1 also binds to CD43 and to Plasmodium falciparum infected RBCs. W-CAM-1 MAb blocks aggregation of cell lines mediated by the ICAM-1 and blocks homotypic binding of purified populations of activated T- and B-lymphocytes and also aggregation of mixed T- and B-cell blasts. It inhibits T-cell adhesion to normal human endothelial cells. Activation induced by cell-cell contact (mixed lymphocyte reaction, T-cell mediated B-cell activation) is significantly inhibited. This MAb blocks elements of both effector arms of immune system (cytotoxic cell function and Ig production).
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 1H4 or Wehi-CAM-1 or W-CAM-1
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC940988-500
- BNC940988-100
- Antigen symbol:
- ICAM1
- ICAM1
- Antigen name:
- CD54
- CD54
- Antigen synonyms:
- Human rhinovirus receptor|ICAM-1|Cell surface glycoprotein P3.58|Ly 47|Intercellular adhesion molecule 1|CD54|MyD10|Surface antigen of activated B cells|Major group rhinovirus receptor|MALA2
- Human rhinovirus receptor|ICAM-1|Cell surface glycoprotein P3.58|Ly 47|Intercellular adhesion molecule 1|CD54|MyD10|Surface antigen of activated B cells|Major group rhinovirus receptor|MALA2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone 1H4 or Wehi-CAM-1 or W-CAM-1
- Clone 1H4 or Wehi-CAM-1 or W-CAM-1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 643447
- 643447
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide