Anti-F13A1 Mouse Monoclonal Antibody [clone: AC-1A1]
Catalog # DIABPDM141
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Coagulation Factor 13 A1
- Antigen symbol:F13A1
- Clonality:Monoclonal
- Clone:AC-1A1
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen synonyms:coagulation factor XIII A chain|A1 polypeptide)|protein-glutamine gamma-glutamyltransferase A chain|A subunit|coagulation factor XIII|coagulation factor XIIIa|fibrinoligase|FSF|bA525O21.1 (coagulation factor XIII|factor XIIIa|TGase|A polypeptide|fibrin stabilizing factor|transglutaminase. plasma|transglutaminase A chain
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: F13A1
Clonality: Monoclonal
Clone: AC-1A1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: