Order Entry
Norway
ContactUsLinkComponent
Anti-CCNA1 0 Monoclonal Antibody [clone: 6E6]
Anti-CCNA1 0 Monoclonal Antibody [clone: 6E6]
Catalog # DIABMOB175
Anti-CCNA1 0 Monoclonal Antibody [clone: 6E6]
Catalog # DIABMOB175

Specifications

  • Antigen name:
    Cyclin A
  • Antigen symbol:
    CCNA1
  • Clonality:
    Monoclonal
  • Clone:
    6E6
  • Antigen synonyms:
    CCN1|CCNA
  • Size:
    1 mL
  • Pk:
    1 ml

Specifications

About this item

Type:
Antigen: CCNA1
Clonality: Monoclonal
Clone: 6000000
Conjugation:
Epitope:
Host: 0
Isotype:
Reactivity: