Anti-C-Myc Mouse Monoclonal Antibody [clone: 9E10]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: C-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Type:
Antigen: C-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: