Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-VIM Rabbit Monoclonal Antibody () [clone: SP20]
Anti-VIM Rabbit Monoclonal Antibody () [clone: SP20]
DIABRMAB019
:
DIAGNOSTIC BIOSYSTEM
Anti-VIM Rabbit Monoclonal Antibody () [clone: SP20]
DIABRMAB019
:
DIAGNOSTIC BIOSYSTEM
Antigen synonyms:
CTRCT30|HEL113
Size:
500 μL
Cat. no.:
DIABRMAB019
Pk:
0,5 ml