Anti-Wilm's Tumor wt1 Mouse Monoclonal Antibody () [clone: 6F-H2]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Wilm's Tumor wt1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: