Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]
Catalog # DIABPDM088
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:S100/ICaBP type calcium binding domain
- Antigen symbol:S100
- Clonality:Monoclonal
- Clone:SH-B1
- Host:Mouse
- Isotype:IgG1
- Antigen synonyms:MGC133384|MYHa|MYHSA1|MyHC-2X/D
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: S100
Clonality: Monoclonal
Clone: SH-B1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: