Anti-Neurofilaments Mouse Monoclonal Antibody [clone: 2F11]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Neurofilaments
Clonality: Monoclonal
Clone: 2F11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: