Anti-Mart-1/Melan-A Mouse Monoclonal Antibody [clone: A103]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Mart-1/Melan-A
Clonality: Monoclonal
Clone: A103
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Frequently Bought Together