Anti-KI67 Mouse Monoclonal Antibody [clone: MB67 (MM1)]
Catalog # DIABPDM026-01
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Monoclonal
- Clone:MB67 (MM1)
- Host:Mouse
- Isotype:IgG1
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: KI67
Clonality: Monoclonal
Clone: MB67 (MM1)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: