Anti-PTPRC Mouse Monoclonal Antibody [clone: PD7/26+2B11]
Catalog # DIABPDM009
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:PD7/26+2B11
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: PTPRC
Clonality: Monoclonal
Clone: PD7/26+2B11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0