Anti-CD79a Mouse Monoclonal Antibody [clone: HM47/A9]
Catalog # DIABPDM125
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Cluster of Differentiation 79a molecule, immunoglobulin-associated alpha
- Antigen symbol:CD79a
- Clonality:Monoclonal
- Clone:HM47/A9
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen synonyms:MB-1|IGA|B-cell antigen receptor complex-associated protein alpha chain|CD79a antigen immunoglobulin-associated alpha|MB-1 membrane glycoprotein
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: CD79a
Clonality: Monoclonal
Clone: HM47/A9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: