Anti-CDK2 Mouse Monoclonal Antibody [clone: 2B6]
Type:
Antigen: CDK2
Clonality: Monoclonal
Clone: 2B6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Type:
Antigen: CDK2
Clonality: Monoclonal
Clone: 2B6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: