Anti-Cd43 T Cell Mouse Monoclonal Antibody [clone: DF-T1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cd43 T Cell
Clonality: Monoclonal
Clone: DF-T1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: