Anti-Cbp Mouse Monoclonal Antibody [clone: AC238]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cbp
Clonality: Monoclonal
Clone: AC238
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Type:
Antigen: Cbp
Clonality: Monoclonal
Clone: AC238
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: