Anti-C1q Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # DIABF010
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen symbol:C1q
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Size:1 mL
- Pk:1 ml
Specifications
About this item
Type:
Antigen: C1q
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0