Anti-TFAP2C Mouse Monoclonal Antibody [clone: A6/2/2]
Leverandør: DIAGNOSTIC BIOSYSTEM
Sertifikat
Om dette produktet
Type:
Antigen: TFAP2C
Clonality: Monoclonal
Clone: A6/2/2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Type:
Antigen: TFAP2C
Clonality: Monoclonal
Clone: A6/2/2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: