Anti-CK14 Mouse Monoclonal Antibody [clone: LL002]
Leverandør: DIAGNOSTIC BIOSYSTEM
Sertifikat
Om dette produktet
This antibody is specific to 50 kDa cytokeratin protein designated cytokeratin 14. Cytokeratin 14 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 15. This antibody can be used to distinguish stratified epithelial cells from simple epithelial cells.
Type:
Antigen: CK14
Clonality: Monoclonal
Clone: LL002
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: