Anti-CCNE1 Mouse Monoclonal Antibody [clone: HE12]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody recognizes a doublet of 50 kDa and a single band of 42 kDa identified as cyclin E. Cyclin proteins are a family of related proteins, which control the progression of the eukaryotic cell cycle. Overexpression of cyclin E shortens the length of the G1 phase, accelerating progression of S phase. The activity of cyclin E is mediated through its activation of cyclin dependent kinase 2 (cdk2) protein and is modulated by the presence of tumor suppressor proteins such as p16.
Type:
Antigen: CCNE1
Clonality: Monoclonal
Clone: HE12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Frequently Bought Together