Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]
Catalog # DIABPDM143
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Fc fragment of IgE receptor 2
- Antigen symbol:FCE2
- Clonality:Monoclonal
- Clone:1B12
- Host:Mouse
- Isotype:IgG
- Antigen synonyms:CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
- Size:6 mL
- Pk:6 ml
Specifications
About this item
This antibody is specific to a membrane glycoprotein of 45 kDa, designated as CD23. CD23 is found on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocytic leukemias.
Type:
Antigen: CD23
Clonality: Monoclonal
Clone: 1B12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: