Anti-ERBB2 Mouse Monoclonal Antibody (Purified) [clone: ERB2/776]
: Biotium
This MAb is specific to c-erbB-2/HER-2 and shows minimal cross-reaction with other members of the family. C-erbB-2/HER-2 is a member of the EGFR family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: ERB2/776
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNUB0776-100
- BNUB0776-500
- Antigen symbol:
- ERBB2
- ERBB2
- Antigen name:
- HER-2 / CD340
- HER-2 / CD340
- Antigen synonyms:
- Verb b2 Erythroblastic Leukemia Viral Oncogene Homolog 2|Neuro/Glioblastoma Derived Oncogene Homolog|CD340|p185
- Verb b2 Erythroblastic Leukemia Viral Oncogene Homolog 2|Neuro/Glioblastoma Derived Oncogene Homolog|CD340|p185
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone ERB2/776
- Clone ERB2/776
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 446352
- 446352
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide