Order Entry
Norway
ContactUsLinkComponent
RNA extraction, MPure™ Total RNA extraction kit , MP Biomedicals
RNA extraction, MPure™ Total RNA extraction kit , MP Biomedicals
Catalog # ICNA117022160
Supplier:  MP Biomedicals
CAS Number:  
RNA extraction, MPure™ Total RNA extraction kit , MP Biomedicals
Catalog # ICNA117022160
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Plant samples
    Blood
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    MPure™ Total RNA Extraction Kit, 48 tests
  • Seq:
    02
  • Description:
    MPure™ Total RNA Extraction Kit, 48 tests

Specifications

About this item

MPure™ Total RNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction of total RNA from whole blood, blood cells, animal tissue, plant tissue, yeast or cultured cells and viral RNA from human biological specimens such as serum, plasma, and other cell-free fluids.