Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Meropenem
Meropenem ≥98%
Meropenem ≥98%
Meropenem ≥98%
Supplier:
Cayman Chemical
Certificates
Expand image
Meropenem ≥98%
Supplier:
Cayman Chemical
About this item
CAS Number:
96036-03-2
:
Formula:
C₁₇H₂₅N₃O₅S
MDL Number:
MFCD00864966
:
383.5 g/mol