Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cod liver oil
Cod liver oil, clear yellow liquid
Cod liver oil, clear yellow liquid
:
MP Biomedicals
Cod liver oil, clear yellow liquid
:
MP Biomedicals
:
8001-69-2
:
:
MFCD00146493
:
Ambient
Identity test:
Passes
Vitamin A:
≥850 USP u/gm
Vitamin D:
≥85 USP u/gm