Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethylvanillin
Ethylvanillin, white powder
Ethylvanillin, white powder
Ethylvanillin, white powder
Catalog # ICNA0215795980
Supplier:
MP Biomedicals
CAS Number:
Expand image
Ethylvanillin, white powder
Catalog # ICNA0215795980
Supplier:
MP Biomedicals
CAS Number:
Specifications
Pk:
100 g
Seq:
3401
Specifications
About this item
Product Details
:
:
166.18 g/mol
:
GHS07
CAS Number:
121-32-4
Formula:
C₉H₁₀O₃
MDL Number:
MFCD00006944
Product Details