Methyl triacontanate, white powder
Katalog # ICNA0215695980
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:466.83 g/mol
Storage Temperature:Ambient
CAS Number:629-83-4
Formula:C₃₁H₆₂O₂
MDL Number:MFCD00042897