kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Propylenkarbonat
Propylenkarbonat, liquid
Propylenkarbonat, liquid
:
MP Biomedicals
Propylenkarbonat, liquid
:
MP Biomedicals
:
108-32-7
:
:
C₄H₆O₃
:
MFCD00005385
:
102.09 g/mol