Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-5-nitroimidazole-1-ethanol
2-Methyl-5-nitroimidazole-1-ethanol, white powder
2-Methyl-5-nitroimidazole-1-ethanol, white powder
2-Methyl-5-nitroimidazole-1-ethanol, white powder
Supplier:
MP Biomedicals
Certificates
Utvid bildet
2-Methyl-5-nitroimidazole-1-ethanol, white powder
Supplier:
MP Biomedicals
Certificates
About this item
CAS Number:
443-48-1
:
Formula:
C₆H₉N₃O₃
MDL Number:
MFCD00009750
:
171.16 g/mol
Melting Pt:
158…160° C (lit.)
Storage Temperature:
Refrigerator
Related Information
Dokumentasjon
Certificates
Search by Lot Number
Dokumentasjon