Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Manganese (II) sulphate monohydrate
Manganese(II) sulphate monohydrate, pale pink powder
Manganese(II) sulphate monohydrate, pale pink powder
Manganese(II) sulphate monohydrate, pale pink powder
Supplier:
MP Biomedicals
Certificates
Expand image
Manganese(II) sulphate monohydrate, pale pink powder
Supplier:
MP Biomedicals
About this item
CAS Number:
10034-96-5
:
Formula:
MnSO₄·1H₂O
MDL Number:
MFCD00149159
ADR:
9,III
:
169.02 g/mol
UN:
3077
:
GHS08
:
GHS09