Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Beclomethasone dipropionate
Beclomethasone dipropionate, white powder
Beclomethasone dipropionate, white powder
Beclomethasone dipropionate, white powder
Supplier:
MP Biomedicals
Certificates
Expand image
Beclomethasone dipropionate, white powder
Supplier:
MP Biomedicals
About this item
CAS Number:
5534-09-8
:
Formula:
C₂₈H₃₇ClO₇
MDL Number:
MFCD00135613
:
521.047 g/mol
:
GHS08
Storage Temperature:
Refrigerator