D-(+)-Trehalose dihydrate ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:6138-23-4
:
Formula:C₁₂H₂₂O₁₁·2H₂O
MDL Number:MFCD00071594
:378.33 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity :#99%
- FTIR:Conforms to Standard
- Loss on Drying :8.5 - 10.5%