Sudan black B, dark powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:4197-25-5
:
Formula:C₂₉H₂₄N₆
MDL Number:MFCD00006919
:456.55 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test :Passes
- UV/Vis: λmax (ethanol):598 ± 4 n